(+/-)-Sulpiride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(+/-)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulphamoylbenzamide
S0501-5G
S0501-25G
TCIAS0501-5GEA
30
EUR
TCIAS0501-5G
TCIAS0501-25G
(+/-)-Sulpiride ≥98.0% (by HPLC, titration analysis)
(+/-)-Sulpiride
Formula:
C₁₅H₂₃N₃O₄S MW: 341,43 g/mol |
Número MDL:
MFCD00055061 Núm. CAS: 15676-16-1 EINECS: 239-753-7 Índice Merck: 14,08989 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 179.0 to 185.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...