Simvastatin ≥97.0% (by HPLC)

Proveedor: TCI

Sinónimos: (1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate

S0509-1G S0509-100MG
TCIAS0509-1GEA 163 EUR
TCIAS0509-1G TCIAS0509-100MG
Simvastatin ≥97.0% (by HPLC)
Simvastatin
Formula: C25H38O5
MW: 418,57 g/mol
Número MDL: MFCD00072007
Núm. CAS: 79902-63-9
EINECS: 404-520-2
Índice Merck: 14,08539

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 132.0 to 137.0 deg-C
Specific rotation [a]20/D +285 to +300 deg (C=1, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR