S-Sodium ethanethiosulphonate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
S0533-25G
S0533-500G
TCIAS0533-25GEA
96
EUR
TCIAS0533-25G
TCIAS0533-500G
S-Sodium ethanethiosulphonate ≥98.0% (by HPLC, titration analysis)
S-Sodium ethanethiosulphonate
Formula:
C₂H₆O₂S₂ |
Núm. CAS:
31999-88-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...