Sodium (trihydroxy)phenylborate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
S0558-5G
S0558-1G
TCIAS0558-5GEA
150
EUR
TCIAS0558-5G
TCIAS0558-1G
Sodium (trihydroxy)phenylborate ≥98.0% (by HPLC, titration analysis)
Sodium (trihydroxy)phenylborate
Formula:
C₆H₈BNaO₃ MW: 161.93 g/mol |
Núm. CAS:
52542-80-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...