Sodium-3-(N-ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulphonate ≥98.0% (by HPLC, titration analysis) for biochemical research
Proveedor: TCI
Sinónimos:
ADOS, 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulphonic acid Sodium Salt, N-Ethyl-N-(2-hydroxy-3-sulphopropyl)-3-methoxyaniline Sodium Salt
S0826-200MG
S0826-1G
TCIAS0826-200MGEA
23
EUR
TCIAS0826-200MG
TCIAS0826-1G
Sodium-3-(N-ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulphonate ≥98.0% (by HPLC, titration analysis) for biochemical research
Sodium-3-(N-ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulphonate
Formula:
C₁₂H₁₉NO₅S MW: 311,33 g/mol Pto. de fusión: 144°C |
Número MDL:
MFCD09261431 Núm. CAS: 82692-96-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Hydrogendioxide detection test (Peroxidase method) | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...