9,9'-Spirobi[9H-fluorene]-2-boronic acid
Proveedor: TCI
|
Atención
|
Sinónimos:
9,9'-Spirobifluorene-2-boronic acid
S0831-1G
S0831-5G
TCIAS0831-1GEA
58
EUR
TCIAS0831-1G
TCIAS0831-5G
9,9'-Spirobi[9H-fluorene]-2-boronic acid
9,9'-Spirobi[9H-fluorene]-2-boronic acid
Formula:
C₂₅H₁₇BO₂ MW: 360,22 g/mol |
Número MDL:
MFCD13248568 Núm. CAS: 236389-21-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 95.0 to 105.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...