Sarafloxacin hydrochloride ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Sarafloxacin HCl, 6-Fluoro-1-(4-fluorophenyl)-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid Hydrochloride
S0840-1G
S0840-5G
TCIAS0840-1GEA
87
EUR
TCIAS0840-1G
TCIAS0840-5G
Sarafloxacin hydrochloride ≥97.0% (by HPLC)
Sarafloxacin hydrochloride
Formula:
C₂₀H₁₇F₂N₃O₃·HCl Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11869789 Núm. CAS: 91296-87-6 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...