Sodium-2-iodobenzenesulphonate ≥97.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Sodium 2-iodobenzenesulphonate, 2-Iodobenzenesulphonic acid Sodium Salt
S0848-1G
S0848-5G
TCIAS0848-1GEA
73
EUR
TCIAS0848-1G
TCIAS0848-5G
Sodium-2-iodobenzenesulphonate ≥97.0% (by HPLC)
Sodium-2-iodobenzenesulphonate
Formula:
C₆H₅IO₃S MW: 306,05 g/mol |
Número MDL:
MFCD12031329 Núm. CAS: 62973-69-7 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Ion exchange titration) | min. 95.0 % (after deducting NaCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...