Sumatriptan succinate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-(3-(2-(Dimethylamino)ethyl)-1H-indol-5-yl)-N-methylmethanesulphonamide succinate, 3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl-N-methylmethanesulphonamide Succinate

S0851-100MG S0851-1G
TCIAS0851-100MGEA 46 EUR
TCIAS0851-100MG TCIAS0851-1G
Sumatriptan succinate ≥98.0% (by HPLC, titration analysis)
Sumatriptan succinate
Formula: C₁₄H₂₁N₃O₂S
MW: 413,49 g/mol
Número MDL: MFCD00902856
Núm. CAS: 103628-48-4
Índice Merck: 14,08997

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR