N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
2,5-Dioxopyrrolidin-1-yl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate
S0866-1G
S0866-200MG
TCIAS0866-1GEA
442
EUR
TCIAS0866-1G
TCIAS0866-200MG
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate
Formula:
C₁₄H₉NO₇ MW: 303,23 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00037568 Núm. CAS: 134471-24-2 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...