Formaldehyde sodium bisulphite hemihydrate ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt
S0880-25G
S0880-500G
TCIAS0880-25GEA
16
EUR
TCIAS0880-25G
TCIAS0880-500G
Formaldehyde sodium bisulphite hemihydrate ≥97.0% (by titrimetric analysis)
Formaldehyde sodium bisulphite hemihydrate
Formula:
CH₄O₄S MW: 134,08 g/mol |
Núm. CAS:
63148-74-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % (calcd.on dried substance) |
Drying loss | 5.5 to 7.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...