Silver bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

S0898-1G S0898-5G
TCIAS0898-1GEA 111 EUR
TCIAS0898-1G TCIAS0898-5G
Silver bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Silver bis(trifluoromethanesulphonyl)imide
Formula: C₂AgF₆NO₄S₂
Temperatura de almacenaje: Congelador
Número MDL: MFCD08457644
Núm. CAS: 189114-61-2
UN: 1759
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Precipitation Titration) min. 98.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR