Sulphadoxin ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: sulphadoxine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulphonamide, 4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulphonamide

S0899-5G S0899-25G
TCIAS0899-5GEA 35 EUR
TCIAS0899-5G TCIAS0899-25G
Sulphadoxin ≥98.0% (by HPLC, titration analysis)
Sulfadoxin
Formula: C₁₂H₁₄N₄O₄S
MW: 310,33 g/mol
Número MDL: MFCD00792890
Núm. CAS: 2447-57-6
EINECS: 219-504-9
Índice Merck: 14,08906
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59e00331596081875c78fcf562856f45ca88280b&vid=59e00331596081875c78fcf562856f45ca88280b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR