Sulphalene ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulphonamide, N-(3-Methoxypyrazin-2-yl)sulphanilamide
S0917-5G
S0917-25G
TCIAS0917-5GEA
49
EUR
TCIAS0917-5G
TCIAS0917-25G
Sulphalene ≥98.0% (by HPLC, titration analysis)
Sulphalene
Formula:
C₁₁H₁₂N₄O₃S MW: 280,30 g/mol |
Número MDL:
MFCD00437754 Núm. CAS: 152-47-6 EINECS: 205-804-7 Índice Merck: 14,08910 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 176.0 to 180.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...