Sodium-tert-pentyloxide ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt

S0950-25G S0950-100G
TCIAS0950-25GEA 16 EUR
TCIAS0950-25G TCIAS0950-100G
Sodium-tert-pentyloxide ≥97.0% (by titrimetric analysis)
Sodium-tert-pentyloxide
Formula: C₅H₁₁NaO
MW: 110,132 g/mol
Número MDL: MFCD00064245
Núm. CAS: 14593-46-5
EINECS: 238-639-4
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=660f8c7fa3c23e28fd44dcf7ed0c90f63c18a782&vid=660f8c7fa3c23e28fd44dcf7ed0c90f63c18a782&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR