Biotin-LC-LC-NHS ≥90.0% (by HPLC)
Proveedor: TCI
Sinónimos:
N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate, Biotin-LC-LC-NHS (2mg*5), 2,5-Dioxopyrrolidin-1-yl 6-(6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanamido)hexanoate, N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic acid N-Succinimidyl ester
S0956-25MG
S0956-100MG
TCIAS0956-25MGEA
98
EUR
TCIAS0956-25MG
TCIAS0956-100MG
Biotin-LC-LC-NHS ≥90.0% (by HPLC)
Biotin-LC-LC-NHS
Formula:
C₂₆H₄₁N₅O₇S MW: 567,702 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00467154 Núm. CAS: 89889-52-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Elemental analysis (Nitrogen) | 11.70 to 13.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...