Sodium pivalate hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Trimethylacetic acid sodium salt hydrate, Sodium trimethylacetate hydrate, Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate

S0978-5G S0978-25G
TCIAS0978-5GEA 38 EUR
TCIAS0978-5G TCIAS0978-25G
Sodium pivalate hydrate ≥98.0% (by titrimetric analysis)
Sodium pivalate hydrate
Formula: C₅H₁₁NaO₃
MW: 142,13 g/mol
Número MDL: MFCD00150782
Núm. CAS: 143174-36-1
EINECS: 214-678-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd518ba35752f3ae2499855659e678997b05e0fd&vid=bd518ba35752f3ae2499855659e678997b05e0fd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 6.5 to 12.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR