Sodium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
S0992-5G
S0992-1G
TCIAS0992-5GEA
262
EUR
TCIAS0992-5G
TCIAS0992-1G
Sodium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
Sodium bis(fluorosulphonyl)imide
Formula:
F₂NNaO₄S₂ MW: 203,11 g/mol |
Número MDL:
MFCD29905041 Núm. CAS: 100669-96-3 UN: 1759 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...