Testosterone ≥98.0% (por GC)

Proveedor: TCI
Peligro

T0027-1G T0027-10G
TCIAT0027-1GEA 23 EUR
TCIAT0027-1G TCIAT0027-10G
Testosterone ≥98.0% (por GC)
Testosterone
Formula: C₁₉H₂₈O₂
Número MDL: MFCD00003654
Núm. CAS: 58-22-0
EINECS: 200-370-5
Índice Merck: 14,09180

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 157.0 deg-C
Specific rotation [a]20/D +99 to +105 deg (C=1, Dioxane)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR