4,5,6,7-Tetrabromophenolphthalein ≥80.0% (by titrimetric analysis)

Proveedor: TCI

T0043-1G T0043-5G
TCIAT0043-1GEA 60 EUR
TCIAT0043-1G TCIAT0043-5G
4,5,6,7-Tetrabromophenolphthalein ≥80.0% (by titrimetric analysis)
4,5,6,7-Tetrabromophenolphthalein
Formula: C₂₀H₁₀Br₄O₄
Número MDL: MFCD00047314
Núm. CAS: 13027-28-6
EINECS: 235-889-6

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Iodometric Titration) min. 80.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR