L-Thyroxine sodium salt pentahydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate pentahydrate, (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic acid sodium salt pentahydrate, Levothyroxine sodium pentahydrate

T0245-1G T0245-100MG
TCIAT0245-1GEA 96 EUR
TCIAT0245-1G TCIAT0245-100MG
L-Thyroxine sodium salt pentahydrate ≥98.0% (by titrimetric analysis)
L-Thyroxine sodium salt pentahydrate
Formula: C₁₅H₁₁I₄NO₄
MW: 798,86 g/mol
Número MDL: MFCD00002596
Núm. CAS: 55-03-8
EINECS: 200-221-4
UN: 2811
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -6.5 to -4.5 deg (C=3, EtOH:1mol/L NaOH=2:1)
Water 8.0 to 12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR