1,3,3-Trimethylindolinobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound

Proveedor: TCI

Sinónimos: 1',3',3'-Trimethylspiro[1(2H)-benzopyran-2,2'-indoline]

T0344-1G T0344-5G
TCIAT0344-1GEA 66 EUR
TCIAT0344-1G TCIAT0344-5G
1,3,3-Trimethylindolinobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound
1,3,3-Trimethylindolinobenzopyrylospiran
Formula: C₁₉H₁₉NO
MW: 277,37 g/mol
Número MDL: MFCD00059174
Núm. CAS: 1485-92-3

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Functionality test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR