Nɑ-Tosyl-L-arginine
Proveedor: TCI
T0684-5G
T0684-1G
TCIAT0684-5GEA
44
EUR
TCIAT0684-5G
TCIAT0684-1G
Nɑ-Tosyl-L-arginine
Nalpha-Tosyl-L-arginine
Formula:
C₁₃H₂₀N₄O₄S |
Número MDL:
MFCD00019732 Núm. CAS: 1159-15-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (after drying) |
Specific rotation [a]20/D | -14.0 to -16.0 deg (C=2, 1mol/L HCl) (after drying) |
Drying loss | 10.0 to 16.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...