Taurocholic acid sodium salt (from bovine bile) ≥84% (by NMR)
Proveedor: TCI
|
Atención
|
Sinónimos:
Cholaic acid sodium salt, Taurocholic acid sodium, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1), Sodium taurocholate
T0808-1G
T0808-25G
T0808-5G
TCIAT0808-1GEA
37
EUR
TCIAT0808-1G
TCIAT0808-25G
TCIAT0808-5G
Taurocholic acid sodium salt (from bovine bile) ≥84% (by NMR)
Taurocholic acid sodium salt
Formula:
C₂₆H₄₄NNaO₇S MW: 537,688 g/mol |
Número MDL:
MFCD00013050 Núm. CAS: 145-42-6 EINECS: 205-653-7 Índice Merck: 14,09075 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (qNMR) | min. 84.0 % |
Specific rotation [a]20/D | +20.0 to +26.0 deg (C=3, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...