Hexaketocyclohexane ≥95.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Hexaoxocyclohexane, Triquinoyl, Cyclohexanehexone
T0876-1G
T0876-5G
TCIAT0876-1GEA
32
EUR
TCIAT0876-1G
TCIAT0876-5G
Hexaketocyclohexane ≥95.0% (by titrimetric analysis)
Hexaketocyclohexane
Formula:
C₆O₆ MW: 168,06 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00001652 Núm. CAS: 527-31-1 EINECS: 208-412-4 |
Resultados test especificaciones
Appearance | White to Brown powder to crystaline |
Purity (Neutralization back titration) | min. 95.0% (calcd.on anh.substace) |
Water | 40.0 to 44.0 % |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...