Ditetradecyl-3,3'-thiodipropionate ≥94.0% (por GC)
Proveedor: TCI
Sinónimos:
3,3'-Thiodipropionic Acid Ditetradecyl Ester, 3,3'-Thiodipropionic Acid Dimyristyl Ester, Dimyristyl 3,3'-Thiodipropionate, 3,3-Thiodipropionicaciddimyristylester
T0911-25G
T0911-500G
TCIAT0911-25GEA
16
EUR
TCIAT0911-25G
TCIAT0911-500G
Ditetradecyl-3,3'-thiodipropionate ≥94.0% (por GC)
Ditetradecyl-3,3'-thiodipropionate
Formula:
C₃₄H₆₆O₄S MW: 570,96 g/mol |
Número MDL:
MFCD00048462 Núm. CAS: 16545-54-3 EINECS: 240-613-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 94.0 % |
Melting point | 49.0 to 54.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...