N,N,N',N'-Tetraacetylethylenediamine ≥98.0% (por GC)
Proveedor: TCI
T0946-25G
T0946-100G
TCIAT0946-25GEA
39
EUR
TCIAT0946-25G
TCIAT0946-100G
N,N,N',N'-Tetraacetylethylenediamine ≥98.0% (por GC)
N,N,N',N'-Tetraacetylethylenediamine
Formula:
C₁₀H₁₆N₂O₄ |
Número MDL:
MFCD00014967 Núm. CAS: 10543-57-4 EINECS: 234-123-8 Índice Merck: 14,09028 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 150.0 to 153.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...