N-Trifluoroacetyl-D-glucosamine ≥96.0% (by total nitrogen basis)
Proveedor: TCI
Sinónimos:
2-Deoxy-2-trifluoroacetamido-D-glucopyranose
T0973-1G
T0973-5G
TCIAT0973-1GEA
194
EUR
TCIAT0973-1G
TCIAT0973-5G
N-Trifluoroacetyl-D-glucosamine ≥96.0% (by total nitrogen basis)
N-Trifluoroacetyl-D-glucosamine
Formula:
C₈H₁₂F₃NO₆ MW: 275,18 g/mol |
Número MDL:
MFCD00059807 Núm. CAS: 36875-26-0 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Specific rotation [a]20/D | +13.0 to +17.0 deg (C=1.5, H2O) (24hrs allow) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...