1,3,5-Triacryloylhexahydro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

T1000-25G T1000-500G T1000-100G
TCIAT1000-25GEA 23 EUR
TCIAT1000-25G TCIAT1000-500G TCIAT1000-100G
1,3,5-Triacryloylhexahydro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
1,3,5-Triacryloylhexahydro-1,3,5-triazine
Formula: C₁₂H₁₅N₃O₃
MW: 249,27 g/mol
Número MDL: MFCD00035738
Núm. CAS: 959-52-4
EINECS: 213-501-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=778d28d3e2def81355ace035ebb2ff4f4eb419ab&vid=778d28d3e2def81355ace035ebb2ff4f4eb419ab&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR