2,2':5',2''-Tertiofeno ≥98.0% (por GC), purified by sublimation

Proveedor: TCI

T1196-1G T1196-5G
TCIAT1196-1GEA 18 EUR
TCIAT1196-1G TCIAT1196-5G
2,2':5',2''-Tertiofeno ≥98.0% (por GC), purified by sublimation
2,2':5',2''-Tertiofeno
Formula: C₁₂H₈S₃
MW: 248,39 g/mol
Número MDL: MFCD00012167
Núm. CAS: 1081-34-1
Índice Merck: 14,09174

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 93.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR