Tetrabutylammonium bifluoride ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

T1295-25G T1295-5G
TCIAT1295-25GEA 394 EUR
TCIAT1295-25G TCIAT1295-5G
Tetrabutylammonium bifluoride ≥95.0% (by titrimetric analysis)
Tetrabutylammonium bifluoride
Formula: C₁₆H₃₇F₂N
Número MDL: MFCD00077877
Núm. CAS: 23868-34-0
UN: 1760
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Colorless to Red to Green clear liquid
Purity (Neutralization titration) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR