Tetramethyldiphosphine disulphide

Proveedor: TCI

Sinónimos: Tetramethylbiphosphine disulphide

T1308-5G T1308-1G
TCIAT1308-5GEA 144 EUR
TCIAT1308-5G TCIAT1308-1G
Tetramethyldiphosphine disulphide
Tetramethyldiphosphine disulphide
Formula: C₄H₁₂P₂S₂
MW: 186.21 g/mol
Número MDL: MFCD00014459
Núm. CAS: 3676-97-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Elemental analysis (Carbon) 25.0 to 26.3 %
Melting point 223.0 to 229.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR