1-(2,4,6-Triisopropylbenzenesulphonyl)imidazole ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-((2,4,6-Triisopropylphenyl)sulphonyl)-1H-imidazole

T1410-25G T1410-5G
TCIAT1410-25GEA 368 EUR
TCIAT1410-25G TCIAT1410-5G
1-(2,4,6-Triisopropylbenzenesulphonyl)imidazole ≥98.0% (by HPLC, titration analysis)
1-(2,4,6-Triisopropylbenzenesulphonyl)imidazole
Formula: C₁₈H₂₆N₂O₂S
MW: 334,48 g/mol
Número MDL: MFCD00005283
Núm. CAS: 50257-40-4
EINECS: 256-509-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR