(S)-2-(4-Methylphenylsulphonamido)-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Tosyl-L-phenylalanine, Tos-Phe-OH
T1443-25G
T1443-5G
TCIAT1443-25GEA
57
EUR
TCIAT1443-25G
TCIAT1443-5G
(S)-2-(4-Methylphenylsulphonamido)-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-(4-Methylphenylsulphonamido)-3-phenylpropanoic acid
Formula:
C₁₆H₁₇NO₄S |
Número MDL:
MFCD00037251 Núm. CAS: 13505-32-3 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 163.0 to 167.0 deg-C |
Specific rotation [a]20/D | -2.5 to -3.0 deg (C=4.5, Acetone) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...