o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1447-1G T1447-10G
TCIAT1447-1GEA 72 EUR
TCIAT1447-1G TCIAT1447-10G
o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
o-Tolualdehyde 2,4-Dinitrophenylhydrazone
Formula: C₁₄H₁₂N₄O₄
MW: 300,27 g/mol
Número MDL: MFCD00191584
Núm. CAS: 1773-44-0
UN: 1325
ADR: 4.1,III

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR