2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
SEM-triphenylphosphonium Chloride, Triphenyl((2-(trimethylsilyl)ethoxy)methyl)phosphonium chloride
T1458-25G
T1458-5G
TCIAT1458-25GEA
322
EUR
TCIAT1458-25G
TCIAT1458-5G
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride
Formula:
C₂₄H₃₀ClOPSi MW: 429,01 g/mol |
Número MDL:
MFCD00043159 Núm. CAS: 82495-75-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...