Diethylisopropylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Diethylisopropylsilyl Triflate, Trifluoromethanesulphonic acid Diethylisopropylsilyl Ester

T1468-1G T1468-5G
TCIAT1468-1GEA 60 EUR
TCIAT1468-1G TCIAT1468-5G
Diethylisopropylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Diethylisopropylsilyl trifluoromethanesulphonate
Formula: C₈H₁₇F₃O₃SSi
MW: 278,36 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00191689
Núm. CAS: 126889-55-2
UN: 1760
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR