(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol ≥98.0% (por GC)
Proveedor: TCI
T1491-1G
T1491-100MG
TCIAT1491-1GEA
353
EUR
TCIAT1491-1G
TCIAT1491-100MG
(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol ≥98.0% (por GC)
(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol
Formula:
C₁₂H₁₆O MW: 176,26 g/mol Pto. Ebullición: 105°C |
Número MDL:
MFCD00066432 Núm. CAS: 34281-92-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Specific rotation [a]20/D | +55.0 to +61.0 deg (C=1, Methanol) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...