(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (por GC) for e.e. determination by NMR
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol, (S)-(+)-alpha-(Trifluoromethyl)-9-anthracenemethanol
T1521-1G
T1521-100MG
TCIAT1521-1GEA
472
EUR
TCIAT1521-1G
TCIAT1521-100MG
(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (por GC) for e.e. determination by NMR
(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol
Formula:
C₁₆H₁₁F₃O MW: 276,26 g/mol |
Número MDL:
MFCD00062968 Núm. CAS: 60646-30-2 EINECS: 262-348-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Optical purity (GC) | min. 99.0 ee% |
Melting point | 134.0 to 136.0 deg-C |
Specific rotation [a]20/D | +27.0 to +34.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...