Tropolone tosylate ≥99.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
7-Oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzenesulphonate, 2-Toluenesulphonyloxytropone, 2-Tosyloxytropone
T1574-25G
T1574-5G
TCIAT1574-25GEA
417
EUR
TCIAT1574-25G
TCIAT1574-5G
Tropolone tosylate ≥99.0% (by GC, titration analysis)
Tropolone tosylate
Formula:
C₁₄H₁₂O₄S |
Número MDL:
MFCD00671574 Núm. CAS: 38768-08-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Ester Value) | min. 99.0 % |
Melting point | 158.0 to 162.0 deg-C |
Solubility in hot Dioxane | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...