Tri-O-acetyl-D-glucal ≥96.0% (por GC)
Proveedor: TCI
Sinónimos:
(-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate
T1596-25G
T1596-5G
TCIAT1596-25GEA
79
EUR
TCIAT1596-25G
TCIAT1596-5G
Tri-O-acetyl-D-glucal ≥96.0% (por GC)
Tri-O-acetyl-D-glucal
Formula:
C₁₂H₁₆O₇ MW: 272,25 g/mol |
Número MDL:
MFCD00063253 Núm. CAS: 2873-29-2 EINECS: 220-709-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 53.0 to 56.0 deg-C |
Specific rotation [a]20/D | -13.0 to -16.0 deg (C=2, EtOH) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...