2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone ≥98.0% (por GC)

Proveedor: TCI
Peligro

Sinónimos: 2-(Trichloroacetyl)pyrrole, 2-(trichloroacetyl)-1h-pyrrole

T1619-25G T1619-5G
TCIAT1619-25GEA 72 EUR
TCIAT1619-25G TCIAT1619-5G
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone ≥98.0% (por GC)
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
Formula: C₆H₄Cl₃NO
MW: 212,46 g/mol
Número MDL: MFCD00128757
Núm. CAS: 35302-72-8
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74fc0076e1650eb24372a0c5bde905cc6afeea7d&vid=74fc0076e1650eb24372a0c5bde905cc6afeea7d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR