Trimethylene bis(4-aminobenzoate) ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1,3-Propanediol Bis(4-aminobenzoate), Propane-1,3-diyl bis(4-aminobenzoate), 1,3-Bis(4-aminobenzoyloxy)propane
T1646-25G
T1646-100G
TCIAT1646-25GEA
44
EUR
TCIAT1646-25G
TCIAT1646-100G
Trimethylene bis(4-aminobenzoate) ≥98.0% (by HPLC, titration analysis)
Trimethylene bis(4-aminobenzoate)
Formula:
C₁₇H₁₈N₂O₄ MW: 314,34 g/mol |
Número MDL:
MFCD00129765 Núm. CAS: 57609-64-0 EINECS: 260-847-9 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...