Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Tris(2,2'-bipyridyl)dichlororuthenium(II), Dichlorotris(2,2'-bipyridyl)ruthenium(II), Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, Tris(2,2-bipyridyl)ruthenium(II) chloride hexahydrate
T1655-1G
T1655-5G
TCIAT1655-1GEA
96
EUR
TCIAT1655-1G
TCIAT1655-5G
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate
Formula:
C₃₀H₃₆Cl₂N₆O₆Ru MW: 748,62 g/mol |
Número MDL:
MFCD00149670 Núm. CAS: 50525-27-4 |
Resultados test especificaciones
Appearance | Yellow to Amber to Dark red powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 12.0 to 15.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...