Dibenzyl-L-tartrate ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
L-Tartaric Acid Dibenzyl Ester
T1671-25G
T1671-5G
TCIAT1671-25GEA
120
EUR
TCIAT1671-25G
TCIAT1671-5G
Dibenzyl-L-tartrate ≥97.0% (por GC)
Dibenzyl-L-tartrate
Formula:
C₁₈H₁₈O₆ MW: 330,34 g/mol |
Número MDL:
MFCD00674554 Núm. CAS: 622-00-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 66.0 to 70.0 deg-C |
Specific rotation [a]20/D | +11.5 to +13.5 deg (C=1, Acetone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...