Tetracaine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate Hydrochloride, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride

T1688-25G T1688-5G
TCIAT1688-25GEA 73 EUR
TCIAT1688-25G TCIAT1688-5G
Tetracaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Tetracaine hydrochloride
Formula: C₁₅H₂₄N₂O₂·HCl
MW: 300,83 g/mol
Número MDL: MFCD00038912
Núm. CAS: 136-47-0
EINECS: 205-248-5
UN: 2811
ADR: 6.1,III
Índice Merck: 14,09188

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 147.0 to 150.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR