2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine ≥98.0% (by GC, total nitrogen)
Proveedor: TCI
|
Atención
|
T1810-5G
T1810-1G
TCIAT1810-5GEA
480
EUR
TCIAT1810-5G
TCIAT1810-1G
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine ≥98.0% (by GC, total nitrogen)
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine
Formula:
C₆Cl₄N₄ |
Número MDL:
MFCD02093495 Núm. CAS: 32980-71-5 EINECS: 251-318-3 |
|
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...