N,N,N',N'-Tetraphenylbenzidine ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: TPB, N,N,N,N-Tetraphenylbenzidine, 4,4'-Bis(diphenylamino)biphenyl

T1812-5G T1812-25G
TCIAT1812-5GEA 132 EUR
TCIAT1812-5G TCIAT1812-25G
N,N,N',N'-Tetraphenylbenzidine ≥98.0% (por GC)
N,N,N',N'-Tetraphenylbenzidine
Formula: C₃₆H₂₈N₂
MW: 488,63 g/mol
Número MDL: MFCD00228123
Núm. CAS: 15546-43-7
EINECS: 239-599-0

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 230.0 to 236.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR