Tri-O-benzyl-D-glucal ≥95.0% (by HPLC)

Proveedor: TCI

Sinónimos: 3,4,6-Tri-O-benzyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tribenzyl Ether

T1859-1G T1859-5G
TCIAT1859-1GEA 74 EUR
TCIAT1859-1G TCIAT1859-5G
Tri-O-benzyl-D-glucal ≥95.0% (by HPLC)
Tri-O-benzyl-D-glucal
Formula: C₂₇H₂₈O₄
MW: 416,52 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00061640
Núm. CAS: 55628-54-1

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 55.0 to 59.0 deg-C
Specific rotation [a]20/D -2.0 to -5.5 deg (C=2.5, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR