(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
(3R,4S,5R,6R)-3,4,5-tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
T1914-1G
T1914-5G
TCIAT1914-1GEA
16
EUR
TCIAT1914-1G
TCIAT1914-5G
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥97.0% (by HPLC)
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol
Formula:
C₃₄H₃₆O₆ MW: 540,65 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00066004 Núm. CAS: 4132-28-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 146.0 to 153.0 deg-C |
Specific rotation [a]20/D | +47.0 to +51.0 deg (C=2, Dioxane) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...